1-Boc-2-methylazetidine-2-carboxylic acid ≥95%

Supplier: Ambeed
Warning

A561369-100MG A561369-250MG
77673-498EA 52 USD
77673-498 77882-353
1-Boc-2-methylazetidine-2-carboxylic acid ≥95%
1-Boc-2-methylazetidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₄
MDL Number: MFCD12406956
CAS Number: 449758-77-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR