Methyl 2,5-dibromo-3-nitrobenzoate ≥98%

Supplier: Ambeed
Danger

A570811-1G A570811-5G A570811-25G
77617-414EA 13.39 USD
77617-414 77749-175 77749-174
Methyl 2,5-dibromo-3-nitrobenzoate ≥98%
Methyl 2,5-dibromo-3-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₅Br₂NO₄
MDL Number: MFCD30802102
CAS Number: 1215298-17-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR