(S)-tert-Butyl 2-(aminomethyl)pyrrolidine-1-carboxylate hydrochloride ≥95%

Supplier: Ambeed
Warning

A560929-1G A560929-250MG
77673-496EA 287 USD
77673-496 77884-381
(S)-tert-Butyl 2-(aminomethyl)pyrrolidine-1-carboxylate hydrochloride ≥95%
(S)-tert-Butyl 2-(aminomethyl)pyrrolidine-1-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₂₁ClN₂O₂
MDL Number: MFCD11101392
CAS Number: 1190890-11-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR