[RuCl(p-cymene)((S)-tolbinap)]Cl ≥98%

Supplier: Ambeed
Warning

A560306-250MG A560306-1G
77617-380EA 57.68 USD
77617-380 77747-153
[RuCl(p-cymene)((S)-tolbinap)]Cl ≥98%
[RuCl(p-cymene)((S)-tolbinap)]Cl

Precaución: for Research Use Only

Formula: C₅₈H₅₄Cl₂P₂Ru
MDL Number: MFCD09753014
CAS Number: 228120-95-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR