Bicyclo[2.2.1]heptane-1-carboxylic acid ≥98%

Supplier: Ambeed
Warning

A560061-100MG A560061-250MG
77884-376EA 152 USD
77884-376 77884-377
Bicyclo[2.2.1]heptane-1-carboxylic acid ≥98%
Bicyclo[2.2.1]heptane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₁₂O₂
MDL Number: MFCD00180984
CAS Number: 18720-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR