AZD-6482 ≥98%

Supplier: Ambeed
Warning

Synonyms: AZD 6482, (-)-2-[[(1R)-1-[7-Methyl-2-(4-morpholinyl)-4-oxo-4H-pyrido[1,2-a]pyrimidin-9-yl]ethyl]amino]benzoic acid, (R)-2-((1-(7-Methyl-2-morpholino-4-oxo-4H-pyrido[1,2-a]pyrimidin-9-yl)ethyl)amino)benzoic acid

A560000-5MG A560000-10MG A560000-50MG
77669-482EA 60 USD
77669-482 77872-936 77872-937
AZD-6482 ≥98%
AZD-6482

Precaución: for Research Use Only

Formula: C₂₂H₂₄N₄O₄
MDL Number: MFCD16659062
CAS Number: 1173900-33-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR