2-Bromo-4,6-dichloropyrimidine ≥95%

Supplier: Ambeed
Warning

A550956-1G A550956-250MG A550956-5G
77893-120EA 108 USD
77893-120 77893-121 77893-122
2-Bromo-4,6-dichloropyrimidine ≥95%
2-Bromo-4,6-dichloropyrimidine

Precaución: for Research Use Only

Formula: C₄HBrCl₂N₂
MDL Number: MFCD22566618
CAS Number: 1399480-88-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR