Alendronate sodium 98% 16-17% H₂O

Supplier: Ambeed
Warning

A550899-5G A550899-100G A550899-25G
77602-334EA 28.7 USD
77602-334 77878-337 77878-338
Alendronate sodium 98% 16-17% H₂O
Alendronate sodium

Precaución: for Research Use Only

Formula: C₄H₁₂NNaO₇P₂
MDL Number: MFCD01861681
CAS Number: 129318-43-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR