tert-Butyl 7-oxo-3,9-diazabicyclo[3.3.1]nonane-3-carboxylate ≥98%

Supplier: Ambeed
Warning

A559623-1G A559623-250MG A559623-5G
77686-439EA 93 USD
77686-439 77901-241 77900-650
tert-Butyl 7-oxo-3,9-diazabicyclo[3.3.1]nonane-3-carboxylate ≥98%
tert-Butyl 7-oxo-3,9-diazabicyclo[3.3.1]nonane-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₀N₂O₃
MDL Number: MFCD23106404
CAS Number: 926659-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR