8-((2S,3R,4R,5S,6R)-6-((((2R,3R,4R)-3,4-Dihydroxy-4-(hydroxymethyl)tetrahydrofuran-2-yl)oxy)methyl)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)-7-hydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one ≥97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Mirificin
A559306-10MG
A559306-5MG
77749-153EA
113
USD
77749-153
77749-154
8-((2S,3R,4R,5S,6R)-6-((((2R,3R,4R)-3,4-Dihydroxy-4-(hydroxymethyl)tetrahydrofuran-2-yl)oxy)methyl)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)-7-hydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one ≥97%
8-((2S,3R,4R,5S,6R)-6-((((2R,3R,4R)-3,4-Dihydroxy-4-(hydroxymethyl)tetrahydrofuran-2-yl)oxy)methyl)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)-7-hydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one
Precaución: for Research Use Only
Formula:
C₂₆H₂₈O₁₃ |
MDL Number:
MFCD29048571 CAS Number: 103654-50-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...