Zinc(II) dibutyldithiocarbamate ≥99%

Supplier: Ambeed
Danger

A550740-25G A550740-100G A550740-1000G
77594-682EA 5.13 USD
77594-682 77602-332 77872-923
Zinc(II) dibutyldithiocarbamate ≥99%
Zinc(II) dibutyldithiocarbamate

Precaución: for Research Use Only

Formula: C₉H₁₉NS₂
MDL Number: MFCD00067274
CAS Number: 136-23-2
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR