Polyethylene glycol 20,000

Supplier: Ambeed
Warning

A550618-1000G A550618-100G A550618-500G
77598-562EA 14.35 USD
77598-562 77598-564 77598-566
Polyethylene glycol 20,000
Polyethylene glycol 20,000

Precaución: for Research Use Only

Formula: H(OCH₂CH₂)nOH
MDL Number: MFCD00081839
CAS Number: 25322-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR