4-Ethynyl-1H-pyrazole ≥98%

Supplier: Ambeed
Warning

A550423-250MG A550423-1G A550423-100MG
77594-680EA 33.83 USD
77594-680 77613-734 77686-411
4-Ethynyl-1H-pyrazole ≥98%
4-Ethynyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₅H₄N₂
MW: 92.099 g/mol
MDL Number: MFCD18207413
CAS Number: 57121-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR