Fluorene-2-boronic acid pinacol ester ≥99%

Supplier: Ambeed
Warning

Synonyms: 9H-Fluorene-2-boronic acid Pinacol ester, 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolane-2-yl)-9H-fluorene, 2-(9H-Fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-(9H-Fluoren-2-yl)-4,4,5,5-tetramethyl-[1,3,2]dioxaborolane

A558444-1G A558444-25G A558444-5G
77901-234EA 17 USD
77901-234 77901-099 77901-794
Fluorene-2-boronic acid pinacol ester ≥99%
Fluorene-2-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₉H₂₁BO₂
MW: 292.18 g/mol
MDL Number: MFCD16294548
CAS Number: 922706-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR