3-((3-Chlorophenoxy)methyl)-1-(tetrahydro-2H-pyran-4-yl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine ≥99%

Supplier: Ambeed
Warning

Synonyms: PF-4800567

A558311-10MG A558311-1MG A558311-5MG
77617-372EA 66.95 USD
77617-372 77617-374 77617-376
3-((3-Chlorophenoxy)methyl)-1-(tetrahydro-2H-pyran-4-yl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine ≥99%
3-((3-Chlorophenoxy)methyl)-1-(tetrahydro-2H-pyran-4-yl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₁₇H₁₈ClN₅O₂
MDL Number: MFCD22123245
CAS Number: 1188296-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR