(2R)-(+)-1,1-Bis(4-methoxyphenyl)-3-methyl-1,2-butanediamine ≥98%

Supplier: Ambeed
Danger

Synonyms: (R)-1,1-Bis(4-methoxyphenyl)-3-methylbutane-1,2-diamine, (R)-DAIPEN

A550033-1G A550033-250MG
77747-128EA 47 USD
77747-128 77747-129
(2R)-(+)-1,1-Bis(4-methoxyphenyl)-3-methyl-1,2-butanediamine ≥98%
(2R)-(+)-1,1-Bis(4-methoxyphenyl)-3-methyl-1,2-butanediamine

Precaución: for Research Use Only

Formula: C₁₉H₂₆N₂O₂
MW: 314.4220 g/mol
MDL Number: MFCD02684539
CAS Number: 166764-19-8
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR