(4-Methoxypyrimidin-5-yl)boronic acid ≥97%

Supplier: Ambeed
Warning

A550008-250MG A550008-100MG
77602-324EA 296.23 USD
77602-324 77872-922
(4-Methoxypyrimidin-5-yl)boronic acid ≥97%
(4-Methoxypyrimidin-5-yl)boronic acid

Precaución: for Research Use Only

Formula: C₅H₇BN₂O₃
MW: 153.93 g/mol
MDL Number: MFCD11865295
CAS Number: 909187-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR