Scyllo inositol ≥98%

Supplier: Ambeed
Warning

Synonyms: rel-(1r,2r,3r,4r,5r,6r)-Cyclohexane-1,2,3,4,5,6-hexaol, Scyllo-Inositol

A549750-100MG A549750-250MG
77872-920EA 110 USD
77872-920 77872-921
Scyllo inositol ≥98%
Scyllo inositol

Precaución: for Research Use Only

Formula: C₆H₁₂O₆
MDL Number: MFCD00065455
CAS Number: 488-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR