1-Cyclopropyl-4-nitro-1H-pyrazole ≥97%
Supplier: Ambeed
|
Warning
|
A549592-1G
A549592-100MG
A549592-250MG
77613-732EA
151.41
USD
77613-732
77891-106
77891-107
1-Cyclopropyl-4-nitro-1H-pyrazole ≥97%
1-Cyclopropyl-4-nitro-1H-pyrazole
Precaución: for Research Use Only
Formula:
C₆H₇N₃O₂ |
MDL Number:
MFCD16810701 CAS Number: 1240568-16-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...