1,4-Diacetoxybenzene ≥98%

Supplier: Ambeed
Warning

Synonyms: Hydroquinone diacetate, 1,4-Phenylene diacetate, p-Phenylene di(acetate)

A549525-25G A549525-5G A549525-500G
77617-348EA 20.6 USD
77617-348 77617-350 77747-127
1,4-Diacetoxybenzene ≥98%
1,4-Diacetoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
MW: 194.19 g/mol
Boiling Pt: 273
MDL Number: MFCD00011643
CAS Number: 1205-91-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR