Securopen ≥98%

Supplier: Ambeed
Danger

Synonyms: Azlocillin sodium salt, D-alpha-([Imidazolidin-2-on-1-yl]carbonylamino)benzylpenicillin, Sodium (2S,5R,6R)-3,3-dimethyl-7-oxo-6-((R)-2-(2-oxoimidazolidine-1-carboxamido)-2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, Azlocillin sodium

A557633-250MG A557633-100MG A557633-1MG A557633-1G
77602-370EA 9.23 USD
77602-370 77627-146 77627-148 77878-351
Securopen ≥98%
Securopen

Precaución: for Research Use Only

Formula: C₂₀H₂₃N₅NaO₆S
MW: 484.483 g/mol
MDL Number: MFCD07793330
CAS Number: 37091-65-9
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR