4-Formyl-3-methylphenylboronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: (4-Formyl-3-methylphenyl)boronic acid

A557347-1G A557347-5G
77884-370EA 264 USD
77884-370 77884-371
4-Formyl-3-methylphenylboronic acid ≥98%
4-Formyl-3-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BO₃
MDL Number: MFCD10566601
CAS Number: 398151-59-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR