3',5-Dihydroxy-4',6,7-trimethoxyflavone ≥98%

Supplier: Ambeed
Danger

Synonyms: 5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-6,7-dimethoxy-4H-chromen-4-one, Eupatorin

A549169-1MG A549169-5MG
77617-344EA 43.26 USD
77617-344 77749-141
3',5-Dihydroxy-4',6,7-trimethoxyflavone ≥98%
3',5-Dihydroxy-4',6,7-trimethoxyflavone

Precaución: for Research Use Only

Formula: C₁₈H₁₆O₇
MW: 344.32 g/mol
MDL Number: MFCD00016929
CAS Number: 855-96-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR