4-Aminopyridine-3-sulfonamide ≥95%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Amino-3-pyridinesulphonamide
A556814-1G
A556814-250MG
77673-484EA
462
USD
77673-484
77673-486
4-Aminopyridine-3-sulfonamide ≥95%
4-Aminopyridine-3-sulfonamide
Precaución: for Research Use Only
Formula:
C₅H₇N₃O₂S MW: 173.19 g/mol |
MDL Number:
MFCD08144072 CAS Number: 75903-62-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...