3-Chloro-5-cyanobenzene-1-sulfonyl chloride ≥98%

Supplier: Ambeed
Danger

A556592-100MG A556592-250MG
77884-367EA 56 USD
77884-367 77884-368
3-Chloro-5-cyanobenzene-1-sulfonyl chloride ≥98%
3-Chloro-5-cyanobenzene-1-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₇H₃Cl₂NO₂S
MDL Number: MFCD18392429
CAS Number: 1131397-77-7
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR