3-Bromo-1-methylpyrazin-2(1H)-one ≥97%

Supplier: Ambeed
Warning

A548464-250MG A548464-1G
77749-139EA 361 USD
77749-139 77749-138
3-Bromo-1-methylpyrazin-2(1H)-one ≥97%
3-Bromo-1-methylpyrazin-2(1H)-one

Precaución: for Research Use Only

Formula: C₅H₅BrN₂O
MW: 189.01 g/mol
MDL Number: MFCD17250183
CAS Number: 1187017-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR