Stachydrine hydrochloride ≥97%

Supplier: Ambeed
Warning

Synonyms: Stachydrine HCl, (S)-2-Carboxy-1,1-dimethylpyrrolidin-1-ium chloride

A556286-1G A556286-25G A556286-5G
77669-470EA 5 USD
77669-470 77872-932 77872-933
Stachydrine hydrochloride ≥97%
Stachydrine hydrochloride
Formula: C₇H₁₃NO₂
MW: 179.64 g/mol
MDL Number: MFCD00060174
CAS Number: 4136-37-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR