5,7-Dihydroxy-2-(4-hydroxyphenyl)-6-((2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)-8-((2S,3R,4S,5S)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)-4H-chromen-4-one ≥98%

Supplier: Ambeed
Warning

Synonyms: Schaftoside

A556269-1MG A556269-5MG
77617-366EA 35.02 USD
77617-366 77747-144
5,7-Dihydroxy-2-(4-hydroxyphenyl)-6-((2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)-8-((2S,3R,4S,5S)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)-4H-chromen-4-one ≥98%
5,7-Dihydroxy-2-(4-hydroxyphenyl)-6-((2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)-8-((2S,3R,4S,5S)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)-4H-chromen-4-one

Precaución: for Research Use Only

Formula: C₂₆H₂₈O₁₄
MW: 564.49 g/mol
MDL Number: MFCD21333256
CAS Number: 51938-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR