Phenanthrene-2-carboxylic acid ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A548367-100MG A548367-250MG
77684-388EA 99 USD
77684-388 77886-364
Phenanthrene-2-carboxylic acid ≥95%, Ambeed.Inc
Phenanthrene-2-carboxylic acid

Precaución: For research use only.

Formula: C₁₅H₁₀O₂
MDL Number: MFCD09865173
CAS Number: 40452-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR