t-Boc-N-Amido-PEG7-Amine ≥98%

Supplier: Ambeed
Warning

Synonyms: O-(2-Aminoethyl)-O′-[2-(Boc-amino)ethyl]hexaethylene glycol, 25-Amino-5,8,11,14,17,20,23-heptaoxa-2-azapentacosanoic acid 1,1-dimethylethyl ester, tert-Butyl (23-amino-3,6,9,12,15,18,21-heptaoxatricosyl)carbamate, BocNH-PEG7-CH2CH2NH2, Boc-PEG-amine (n=7), Boc-NH-PEG7-NH2

A556004-100MG A556004-250MG A556004-5G
77617-362EA 44.29 USD
77617-362 77686-429 77686-430
t-Boc-N-Amido-PEG7-Amine ≥98%
t-Boc-N-Amido-PEG7-Amine

Precaución: for Research Use Only

Formula: C₂₁H₄₄N₂O₉
MW: 468.582 g/mol
MDL Number: MFCD03453229
CAS Number: 206265-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR