(3-Amino-5-Carbamoylphenyl)boronic acid ≥98%

Supplier: Ambeed
Warning

A555900-1G A555900-250MG
77901-315EA 207 USD
77901-315 77901-854
(3-Amino-5-Carbamoylphenyl)boronic acid ≥98%
(3-Amino-5-Carbamoylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₉BN₂O₃
MW: 179.97 g/mol
MDL Number: MFCD22192442
CAS Number: 1801408-17-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR