2,6-Difluoro-4-carboxyphenylboronic acid ≥98%

Supplier: Ambeed
Warning

A548003-100MG A548003-1G A548003-5G
77884-352EA 46 USD
77884-352 77884-353 77884-354
2,6-Difluoro-4-carboxyphenylboronic acid ≥98%
2,6-Difluoro-4-carboxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BF₂O₄
MDL Number: MFCD17214253
CAS Number: 1031857-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR