4-Bromo-2,3,5,6-tetrafluorobenzoic acid ≥98%

Supplier: Ambeed
Warning

A555879-1G A555879-5G A555879-10G A555879-25G
77673-480EA 45 USD
77673-480 77673-482 77882-335 77882-336
4-Bromo-2,3,5,6-tetrafluorobenzoic acid ≥98%
4-Bromo-2,3,5,6-tetrafluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇HBrF₄O₂
MDL Number: MFCD00075244
CAS Number: 4707-24-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR