2-(1-((2-(Methylamino)pyrimidin-5-yl)methyl)piperidin-3-yl)-6-(thiophen-2-yl)pyrimidin-4(3H)-one ≥99%

Supplier: Ambeed
Warning

Synonyms: Ribocil

A555623-10MG A555623-2MG A555623-5MG
77747-141EA 188 USD
77747-141 77747-142 77747-143
2-(1-((2-(Methylamino)pyrimidin-5-yl)methyl)piperidin-3-yl)-6-(thiophen-2-yl)pyrimidin-4(3H)-one ≥99%
2-(1-((2-(Methylamino)pyrimidin-5-yl)methyl)piperidin-3-yl)-6-(thiophen-2-yl)pyrimidin-4(3H)-one

Precaución: for Research Use Only

Formula: C₁₉H₂₂N₆OS
MW: 382.48 g/mol
CAS Number: 1381289-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR