6-Fluorophthalazin-1(2H)-one ≥97%

Supplier: Ambeed
Warning

A547408-100MG A547408-1G A547408-250MG
77749-135EA 32 USD
77749-135 77749-136 77749-137
6-Fluorophthalazin-1(2H)-one ≥97%
6-Fluorophthalazin-1(2H)-one

Precaución: for Research Use Only

Formula: C₈H₅FN₂O
MDL Number: MFCD22394247
CAS Number: 23928-51-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR