5-Methoxy-1-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-c]pyridine ≥98%

Supplier: Ambeed
Warning

A554981-100MG A554981-1G A554981-250MG
77749-145EA 162 USD
77749-145 77749-146 77749-147
5-Methoxy-1-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-c]pyridine ≥98%
5-Methoxy-1-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-c]pyridine

Precaución: for Research Use Only

Formula: C₁₅H₂₁BN₂O₃
MW: 288.15 g/mol
MDL Number: MFCD16659110
CAS Number: 1310705-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR