3-(2,4-Dimethoxyphenyl)-1-(4-hydroxyphenyl)propan-1-one ≥98%

Supplier: Ambeed
Warning

Synonyms: Loureirin A

A547369-1MG A547369-5MG A547369-10MG
77617-334EA 17.51 USD
77617-334 77617-336 77747-117
3-(2,4-Dimethoxyphenyl)-1-(4-hydroxyphenyl)propan-1-one ≥98%
3-(2,4-Dimethoxyphenyl)-1-(4-hydroxyphenyl)propan-1-one

Precaución: for Research Use Only

Formula: C₁₇H₁₈O₄
MW: 286.32 g/mol
MDL Number: MFCD10566607
CAS Number: 119425-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR