6-(4-(Pyridin-4-ylamino)phenyl)-4,5-dihydropyridazin-3(2H)-one hydrochloride ≥97%

Supplier: Ambeed
Warning

Synonyms: Senazodan HCl

A547109-1MG A547109-10MG A547109-5MG
77617-332EA 27.81 USD
77617-332 77747-115 77747-116
6-(4-(Pyridin-4-ylamino)phenyl)-4,5-dihydropyridazin-3(2H)-one hydrochloride ≥97%
6-(4-(Pyridin-4-ylamino)phenyl)-4,5-dihydropyridazin-3(2H)-one hydrochloride

Precaución: for Research Use Only

Formula: C₁₅H₁₅ClN₄O
MW: 302.76 g/mol
CAS Number: 98326-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR