3-Bromo-2-fluoro-5-(trifluoromethyl)pyridine ≥95%

Supplier: Ambeed
Danger

A547039-1G A547039-5G A547039-250MG
77594-672EA 47.15 USD
77594-672 77602-310 77872-916
3-Bromo-2-fluoro-5-(trifluoromethyl)pyridine ≥95%
3-Bromo-2-fluoro-5-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₂BrF₄N
MW: 243.987 g/mol
MDL Number: MFCD08061304
CAS Number: 1031929-01-7
UN: 1992
ADR: 3,6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR