(S)-(-)-3-Bromo-α-methylbenzylamine ≥98%

Supplier: Ambeed
Danger

Synonyms: (S)-1-(3-Bromophenyl)ethylamine, (S)-1-(3-Bromophenyl)ethanamine

A554454-10G A554454-1G A554454-5G
77686-423EA 127 USD
77686-423 77686-424 77686-425
(S)-(-)-3-Bromo-α-methylbenzylamine ≥98%
(S)-(-)-3-Bromo-α-methylbenzylamine

Precaución: for Research Use Only

Formula: C₈H₁₀BrN
MW: 200.08 g/mol
MDL Number: MFCD00671638
CAS Number: 139305-96-7
UN: 2735
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR