3,5,3',4'-Tetrahydroxy-7-methoxyflavone ≥95%

Supplier: Ambeed
Warning

Synonyms: 2-(3,4-Dihydroxyphenyl)-3,5-dihydroxy-7-methoxy-4H-chromen-4-one, Rhamnetin

A554234-1MG A554234-5MG
77602-350EA 63.55 USD
77602-350 77669-450
3,5,3',4'-Tetrahydroxy-7-methoxyflavone ≥95%
3,5,3',4'-Tetrahydroxy-7-methoxyflavone

Precaución: for Research Use Only

Formula: C₁₆H₁₂O₇
MW: 316.27 g/mol
MDL Number: MFCD00016931
CAS Number: 90-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR