6-Chloro-4-nitro-1H-indole ≥97%

Supplier: Ambeed
Warning

A554210-100MG A554210-250MG
77878-343EA 15 USD
77878-343 77878-344
6-Chloro-4-nitro-1H-indole ≥97%
6-Chloro-4-nitro-1H-indole

Precaución: for Research Use Only

Formula: C₈H₅ClN₂O₂
MDL Number: MFCD07781454
CAS Number: 245524-95-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR