MK-886 ≥98%

Supplier: Ambeed
Warning

Synonyms: 3-(3-(tert-Butylthio)-1-(4-chlorobenzyl)-5-isopropyl-1H-indol-2-yl)-2,2-dimethylpropanoic acid, 3-[1-(p-Chlorobenzyl)-5-(isopropyl)-3-t-butylthioindol-2-yl]-2,2-dimethylpropanoic acid Na, L-663,536

A546613-10MG A546613-1MG A546613-5MG
77613-724EA 36.05 USD
77613-724 77613-726 77613-728
MK-886 ≥98%
MK-886

Precaución: for Research Use Only

Formula: C₂₇H₃₃ClNO₂S·Na
MW: 494.0 g/mol
MDL Number: MFCD00876710
CAS Number: 118414-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR