Thioacetazone ≥98%

Supplier: Ambeed
Danger

Synonyms: N-(4-((2-Carbamothioylhydrazono)methyl)phenyl)acetamide, N-[4-[(2-Carbamothioylhydrazono)methyl]phenyl]acetamide, Thiacetazone, 4-Acetylaminobenzaldehyde thiosemicarbazone, (4-Acetylamino)benzaldehyde Thiosemicarbazone

A546459-1G A546459-250MG
77627-124EA 61.8 USD
77627-124 77627-126
Thioacetazone ≥98%
Thioacetazone

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₄OS
MW: 236.2935 g/mol
MDL Number: MFCD00022157
CAS Number: 104-06-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR