Diethylcarbamazine citrate ≥98%

Supplier: Ambeed
Danger

A553770-1G A553770-100MG
77602-344EA 12.3 USD
77602-344 77627-140
Diethylcarbamazine citrate ≥98%
Diethylcarbamazine citrate

Precaución: for Research Use Only

Formula: C₁₆H₂₉N₃O₈
MDL Number: MFCD00039124
CAS Number: 1642-54-2
UN: 2811
ADR: 6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR