1-Boc-7-(methoxycarbonyl)indole-2-boronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: (1-(tert-Butoxycarbonyl)-7-(methoxycarbonyl)-1H-indol-2-yl)boronic acid

A552779-1G A552779-250MG A552779-5G
77884-361EA 128 USD
77884-361 77884-362 77884-363
1-Boc-7-(methoxycarbonyl)indole-2-boronic acid ≥98%
1-Boc-7-(methoxycarbonyl)indole-2-boronic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₈BNO₆
MW: 319.12 g/mol
MDL Number: MFCD11504860
CAS Number: 953411-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR