5-Carbamoyl-2-fluorophenylboronic acid ≥98%

Supplier: Ambeed
Warning

A552537-1G A552537-5G
77884-359EA 150 USD
77884-359 77884-360
5-Carbamoyl-2-fluorophenylboronic acid ≥98%
5-Carbamoyl-2-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BFNO₃
MDL Number: MFCD08436055
CAS Number: 874289-39-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR