4-Aminopyridine-2,6-dicarboxylic acid ≥95%

Supplier: Ambeed
Warning

A544894-100MG A544894-1G
77886-359EA 139 USD
77886-359 77886-360
4-Aminopyridine-2,6-dicarboxylic acid ≥95%
4-Aminopyridine-2,6-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₇H₆N₂O₄
MDL Number: MFCD08272117
CAS Number: 2683-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR