N,N-Dimethylpiperazine-1-sulfonamide ≥95%

Supplier: Ambeed
Warning

Synonyms: 1-(N,N-Dimethylsulphamoyl)piperazine

A544778-1G A544778-5G
77880-179EA 86 USD
77880-179 77880-180
N,N-Dimethylpiperazine-1-sulfonamide ≥95%
N,N-Dimethylpiperazine-1-sulfonamide

Precaución: for Research Use Only

Formula: C₆H₁₅N₃O₂S
MDL Number: MFCD04972523
CAS Number: 98961-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR