3,5-Dichloro-4-hydroxyphenylboronic acid ≥95%

Supplier: Ambeed
Warning

A551648-100MG A551648-250MG
77900-535EA 50 USD
77900-535 77901-579
3,5-Dichloro-4-hydroxyphenylboronic acid ≥95%
3,5-Dichloro-4-hydroxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BCl₂O₃
MDL Number: MFCD19105364
CAS Number: 1335048-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR